Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000494224
Encompass_Preferred
11 5 mm OD BTO premium high-temp center guide GC septa PK50
Catalog No. 50-004-94224 Supplier Trajan Scientific Supplier No. 298777
11 5 mm OD BTO premium high-temp center guide GC septa PK50