Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000494189
Encompass_Preferred
11 mm OD BTO premium high-temp center guide GC septa PK50
Catalog No. 50-004-94189 Supplier Trajan Scientific Supplier No. 298717
11 mm OD BTO premium high-temp center guide GC septa PK50