Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000494632
Encompass_Preferred
10 mm OD BTO premium high-temp GC septa PK50
Catalog No. 50-004-94632 Supplier Trajan Scientific Supplier No. 298745
10 mm OD BTO premium high-temp GC septa PK50