Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000494014
Encompass_Preferred
9 5 mm (3/8 ) OD BTO premium high-temp septa PK50
Catalog No. 50-004-94014 Supplier Trajan Scientific Supplier No. 298705
9 5 mm (3/8 ) OD BTO premium high-temp septa PK50