Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000493882
Encompass_Preferred
17 mm OD BTO premium high-temp center guide GC septa PK50
Catalog No. 50-004-93882 Supplier Trajan Scientific Supplier No. 298817
17 mm OD BTO premium high-temp center guide GC septa PK50