Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Catalog No. 5000494042
Encompass_Preferred
12 7 mm OD BTO premium high-temp center guide GC septa PK50
Catalog No. 50-004-94042 Supplier Trajan Scientific Supplier No. 298727
12 7 mm OD BTO premium high-temp center guide GC septa PK50