Learn More
Trajan Scientific Septa Inlet General Blue 100/PK
SDP
Catalog No. 5000493851
Encompass_Preferred
11 mm OD General purpose blue GC septa PK100
Catalog No. 50-004-93851 Supplier Trajan Scientific Supplier No. 234115
11 mm OD General purpose blue GC septa PK100