Learn More
Trajan Scientific Septa Inlet General Blue 100/PK
SDP
Catalog No. 5000494930
Encompass_Preferred
12 7 mm (1/2 ) OD General purpose blue GC septa PK100
Catalog No. 50-004-94930 Supplier Trajan Scientific Supplier No. 234125
12 7 mm (1/2 ) OD General purpose blue GC septa PK100