Learn More
Trajan Scientific Septa Inlet General Blue 100/PK
SDP
Catalog No. 5000494214
Encompass_Preferred
9 mm OD General purpose blue GC septa PK100
Catalog No. 50-004-94214 Supplier Trajan Scientific Supplier No. 234109
9 mm OD General purpose blue GC septa PK100