Learn More
Trajan Scientific Septa Inlet General Blue 50/PK
SDP
Catalog No. 5000494129
Encompass_Preferred
12 7 mm (1/2 ) OD General purpose blue GC septa PK50
This item is not returnable. View return policy
Catalog No. 50-004-94129 Supplier Trajan Scientific Supplier No. 234126
12 7 mm (1/2 ) OD General purpose blue GC septa PK50