Learn More
Trajan Scientific Septa Inlet Marathon Long Life 1/EA
SDP
Catalog No. 5000494089
Encompass_Preferred
11 5 mm OD Marathon long-life center guide GC septa PK25
Catalog No. 50-004-94089 Supplier Trajan Scientific Supplier No. 239787
11 5 mm OD Marathon long-life center guide GC septa PK25