Learn More
Trajan Scientific Septa Inlet Marathon Long Life 24/PK
SDP
Catalog No. 5000493983
Encompass_Preferred
17 mm OD Marathon long-life center guide GC septa PK24
This item is not returnable. View return policy
Catalog No. 50-004-93983 Supplier Trajan Scientific Supplier No. 041910
17 mm OD Marathon long-life center guide GC septa PK24