Learn More
Trajan Scientific Septa Inlet Marathon Long Life 25/PK
SDP
Catalog No. 5000494535
Encompass_Preferred
11 5 mm OD Marathon long-life center guide GC septa PK25
This item is not returnable. View return policy
Catalog No. 50-004-94535 Supplier Trajan Scientific Supplier No. 041907
11 5 mm OD Marathon long-life center guide GC septa PK25