Learn More
Trajan Scientific Septa Inlet Marathon Long Life 25/PK
SDP
Catalog No. 5000494208
Encompass_Preferred
9 mm OD Marathon long-life center guide GC septa PK25
Catalog No. 50-004-94208 Supplier Trajan Scientific Supplier No. 239778
9 mm OD Marathon long-life center guide GC septa PK25