Learn More
Trajan Scientific Septa Inlet Marathon Long Life 50/PK
SDP
Catalog No. 5000493782
Encompass_Preferred
17 mm OD Marathon long-life center guide GC septa PK50
This item is not returnable. View return policy
Catalog No. 50-004-93782 Supplier Trajan Scientific Supplier No. 239698
17 mm OD Marathon long-life center guide GC septa PK50