Learn More
Trajan Scientific Septa Inlet Marathon Long Life 50/PK
SDP
Catalog No. 5000494206
Encompass_Preferred
9 mm OD Marathon long-life center guide GC septa PK50
Catalog No. 50-004-94206 Supplier Trajan Scientific Supplier No. 239779
9 mm OD Marathon long-life center guide GC septa PK50