Learn More
Trajan Scientific Septa Inlet Puresep coated 50/PK
SDP
Catalog No. 5000494902
Encompass_Preferred
11 mm OD Puresep T coated GC septa PK50
Catalog No. 50-004-94902 Supplier Trajan Scientific Supplier No. 230685
11 mm OD Puresep T coated GC septa PK50