Learn More
Trajan Scientific Septa Inlet Puresep coated 50/PK
SDP
Catalog No. 5000493601
Encompass_Preferred
9 5 mm (3/8 ) OD Puresep T coated GC septa PK50
Catalog No. 50-004-93601 Supplier Trajan Scientific Supplier No. 230675
9 5 mm (3/8 ) OD Puresep T coated GC septa PK50