Learn More
Chemglass Life Sciences Installation Tool for Septa
SDP
Catalog No. 50143886
Encompass_Preferred
Installation Tool for Septa for CG4814 series of Mini Inert Valve.
Catalog No. 50-143-886 Supplier Chemglass Life Sciences Supplier No. CG4914002
Installation Tool for Septa for CG4814 series of Mini Inert Valve.