Learn More
Trajan Scientific La-Pha-Pack , Septa, 10mm, For use With 10-425 Threaded Caps, PTFE/Silicone, 100 Pack. Made in Germany.
Catalog No. 50711782
Encompass_Preferred
La-Pha-Pack , Septa, 10mm, For use With 10-425 Threaded Caps, PTFE/Silicone, 100 Pack. Made in Germany.
Catalog No. 50-711-782 Supplier Trajan Scientific Supplier No. LAP.09021260
La-Pha-Pack , Septa, 10mm, For use With 10-425 Threaded Caps, PTFE/Silicone, 100 Pack. Made in Germany.