Learn More
Waters Corp Septum, PTFE/Silicone, 7 mm, 100/pk
Supplier:  Waters Corp WAT058874
Encompass_Preferred
The PTFE/Silicone Septum provides a protective barrier for samples during preparation protocols, ensuring cleanliness and dimensional tolerances. It facilitates successful sample injection by allowing a needle to pierce and enter the vial without contamination, actively preventing issues such as leaching and sticking.
  • Acts as a barrier between the sample vial and the outside environment.
  • Allows a needle to pierce and enter the vial without added considerations for sample extraction.
  • Actively prevents leaching and sticking.
  • Held to a high standard of cleanliness and dimensional tolerances.
Catalog No. 50-785-840