Learn More
Ace Glass, Inc. Septa, gray silicone, 3mm x 13mm OD, fits #11 Ace-Thred, pk/12
SDP
Catalog No. 50983477
Encompass
Septa, gray silicone, 3mm x 13mm OD, fits #11 Ace-Thred, pk/12
Catalog No. 50-983-477 Supplier Ace Glass, Inc. Supplier No. 1290410
Septa, gray silicone, 3mm x 13mm OD, fits #11 Ace-Thred, pk/12