Learn More
Waters Corp Waters Corporation Septum, PTFE/Silicone, 100/pk
Catalog No. 50786184 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters offers a selection of cap, septum, and assembled cap/septum for various vial sizes. The PTFE/Silicone Septum is recommended for sample preparation applications, as it allows for sample injection without adding exterior contaminants. A septum protects your sample from external contamination by acting as a barrier between your sample vial and the outside world.
  • Prevents exterior contaminants from entering the vial
  • Simple and easy to use with a sample injector needle
  • Allows maximum content retrieval
  • Supplied in a pack of 100
  • Strictly tested for cleanliness and proper use
Catalog No. 50-786-184 Supplier Waters Corp Supplier No. WAT210685
Waters offers a selection of cap, septum, and assembled cap/septum for various vial sizes. The PTFE/Silicone Septum is recommended for sample preparation applications, as it allows for sample injection without adding exterior contaminants. A septum protects your sample from external contamination by acting as a barrier between your sample vial and the outside world.
  • Prevents exterior contaminants from entering the vial
  • Simple and easy to use with a sample injector needle
  • Allows maximum content retrieval
  • Supplied in a pack of 100
  • Strictly tested for cleanliness and proper use