Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Rid-X Septic System Treatment, Concentrated Powder, Unscented, 9.8 oz, 12/Carton

Catalog No. 502870194
Encompass

Rid-X Septic System Treatment is a concentrated powder designed for maintaining septic systems and drains. It contains a blend of bacteria and enzymes to help break down waste.

  • Form: Powder
  • Container type: Box
  • Container size: 9.8 oz
  • Scent: Unscented
  • For use with septic tanks and pipes

Catalog No. 50-287-0194 Supplier MSC Supplier No. 91510743
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rid-X Septic System Treatment is a concentrated powder designed for maintaining septic systems and drains. It contains a blend of bacteria and enzymes to help break down waste.

  • Form: Powder
  • Container type: Box
  • Container size: 9.8 oz
  • Scent: Unscented
  • For use with septic tanks and pipes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.