Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Septihol™ Ethyl WFI Sterile Alcohol Solution

Catalog No. 19415118 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
22 oz.
32 oz.
1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-118 22 oz.
19-415-119 32 oz.
19-415-120 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-415-118 Supplier STERIS Supplier No. 1S1177
Add to Cart
Add to Cart
This item is not returnable. View return policy

A solution of 70% (v/v) reagent-grade alcohol and 30% (v/v) USP WFI designed to remove organic and inorganic residues in aseptic processing areas; double bagged for easy entry into critical environments.

  • Designed to remove organic and inorganic residue from product, and non-product contact surfaces
  • A mixture of ethanol, isopropanol, methanol and United States Pharmacopeia (USP) Water For Injection (WFI)
  • Free of stabilizers and non-volatile denaturants for quick and complete evaporation
  • Ready to use, sterile alcohol eliminates in-house production validation and documentation concerns
  • Packages are double bagged for easy entry into controlled environments and aseptic core areas
  • Includes extensive documentation package to meet validation objectives
  • Each lot documented by a Certificate of Analysis including irradiation and sterility results
  • Supported by the STERIS Technical Services team for individualized application and validation assistance

Specifications

Product Type Ethyl Alcohol
Sterility Sterile
Composition 70% (v/v) Reagent-Grade Alcohol, and 30% (v/v) USP Water for Injection (WFI)
Filtered Filtered
For Use With (Application) Removing Organic and Inorganic Residues in Aseptic Processing Areas
Quantity 12/Cs.
Volume (English) 22 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.