Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Septihol™ Ethyl WFI Non-Sterile Alcohol Solution

Catalog No. 19415159 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-159 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-415-159 Supplier STERIS Supplier No. 625608WR

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Ethyl WFI Alcohol Solution is designed to remove organic and inorganic residues in aseptic processing areas.

  • Septihol™ Ethyl WFI Alcohol Solution
  • Low residue ethanol based solution
  • Non-sterile
  • Quality
    • Extremely low level of endotoxins from WFI produced by a Finn-Aqua™ Multiple Effect Water Still.
  • Productivity
    • Ready to use alcohol eliminates in-house production validation and documentation concerns.
  • Assurance
    • Each lot documented by a Certificate of Analysis

Specifications

Product Type Alcohol Solution
pH 4.7
Sterility Non-sterile
Container Type Bottle
For Use With (Application) Sanitizer Cleanroom, Controlled Environments
Volume (English) 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.