Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Septihol™ Sterile Alcohol Solution

Catalog No. 19415111 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
16 oz.
32 oz.
1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-415-111 16 oz.
19-415-112 32 oz.
19-415-113 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-415-111 Supplier STERIS Supplier No. 1S35Y4
Add to Cart
Add to Cart

A ready to use 70% (v/v) USP-grade sterile isopropyl alcohol solution that kills a range of Gram-positive and Gram-negative bacteria and has fungicidal activity against Trichophyton interdigitale. Double bagged and gamma irradiated for use in aseptic areas.

  • Ideal as a final microbial control and disinfection step to remove residue from surfaces and to decontaminate gloves
  • EPA biocidal and fungicidal claims are proven in the presence of 5% organic soil on hard, non-porous surfaces
  • Can be used straight out of the container, poured or sprayed
  • Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
  • Free of stabilizers and denaturants for quick and complete evaporation
  • Double-bagged and gamma irradiated for use in aseptic areas
  • Each lot is documented with a Certificate of Analysis for lot traceability

Specifications

Product Type Alcohol Solution
Sterility Sterile
Composition 70% (v/v) USP-Grade Sterile Isopropyl Alcohol
Container Type Trigger Spray Bottle
Format RTU
For Use With (Application) Removing Residue From Surfaces, Decontaminating Gloves
Quantity 12/Cs.
Volume (English) 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.