Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Ideal as a final microbial control and disinfection step to remove residue from surfaces and to decontaminate gloves
- EPA biocidal and fungicidal claims are proven in the presence of 5% organic soil on hard, non-porous surfaces
- Can be used straight out of the container, poured or sprayed
- Ready to use sterile alcohol eliminates in-house production validation and documentation concerns
- Free of stabilizers and denaturants for quick and complete evaporation
- Double-bagged and gamma irradiated for use in aseptic areas
- Each lot is documented with a Certificate of Analysis for lot traceability
Specifications
Specifications
| Product Type | Alcohol Solution |
| Sterility | Sterile |
| Composition | 70% (v/v) USP-Grade Sterile Isopropyl Alcohol |
| Format | RTU |
| For Use With (Application) | Removing Residue From Surfaces, Decontaminating Gloves |
| Quantity | 12/Cs. |
| Volume (English) | 32 oz. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?