Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Septihol WFI Sterile Alcohol Wipes

Catalog No. 17100902 Shop All STERIS Corporation Products
Change view
Click to view available options
Length (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
17-100-902 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-902 Supplier STERIS Supplier No. 624915
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile alcohol wipes designed for cleanroom use, offering a convenient and low-particulate solution for controlled environments.

Septihol WFI Sterile Alcohol Wipes are part of the Septihol family of products from STERIS. Each product is designed to amplify your validation requirements while providing multiple packaging options.

  • 70% (v/v) USP isopropanol and 30% USP Water For Injection (WFI) alcohol solution
  • Specifically formulated for easy evaporation
  • Right-sized packaging
  • Knitted white polyester with sealed edges
  • Double-bagged and gamma irradiated, with every lot USP sterility tested
  • Low endotoxin tested

Benefits

  • Industry best practice for cleaning and rinsing
  • Ideal for use on critical surfaces
  • Reduced waste of wipes
  • Low particulate wipes to achieve compliance with current annex 1 contamination control strategies
  • Facilitates introduction into critical areas

Specifications

Product Type IPA Wipe
Length (English) 12 in.
Length (Metric) 30.48 cm
Width (English) 12 in.
Width (Metric) 30.48 cm
Material Polyester
Dimensions (L x W) 12 x 12 in.
Packaging Quantity 12 Pk. x 15 wipes per case
Saturation Solution 70% IPA/30% USP WFI
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.