Learn More
ATCC Septoria tritici
Catalog No. 502381526
Encompass
Major wheat pathogen that causes Septoria tritici blotch. This strain was isolated in 1970 from wheat (Triticum aestivum) in Minnesota, United States. This product is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant.
Catalog No. 50-238-1526 Supplier ATCC Supplier No. 26517
Major wheat pathogen that causes Septoria tritici blotch. This strain was isolated in 1970 from wheat (Triticum aestivum) in Minnesota, United States. This product is provided as an ampoule containing viable cells (yeast cells, spores, or agar cubes with mycelia) suspended in cryoprotectant.