Learn More
Agilent Technologies Inlet Septa, Bleed and Temperature Optimized, Silicone, 11 mm, 100/Pk.
Catalog No. NC1443158
Encompass
These 11 mm bleed and temperature optimized (BTO) inlet septa are injection-molded from high-quality silicone for uniform dimensions. Designed for use in GC and GC/MS applications, they feature a proprietary non-stick plasma treatment that minimizes background signal and reduces septum sticking in the injection port. The integrated centering guide helps ensure consistent needle puncture placement, improving resealability and extending septum life.
  • Features a maximum operating temperature of 400°C.
  • Plasma-treated surface eliminates contaminants to reduce ghost peaks.
  • Compatible with split/splitless (S/SL), programmable temperature vaporizer (PTV), and multi-mode injection (MMI) inlets.
  • Supplied clean and ready to use in individually sealed packaging.
  • Highly recommended for trace analysis.
Catalog No. NC1443158 Supplier Agilent Technologies Supplier No. 51834757100
These 11 mm bleed and temperature optimized (BTO) inlet septa are injection-molded from high-quality silicone for uniform dimensions. Designed for use in GC and GC/MS applications, they feature a proprietary non-stick plasma treatment that minimizes background signal and reduces septum sticking in the injection port. The integrated centering guide helps ensure consistent needle puncture placement, improving resealability and extending septum life.
  • Features a maximum operating temperature of 400°C.
  • Plasma-treated surface eliminates contaminants to reduce ghost peaks.
  • Compatible with split/splitless (S/SL), programmable temperature vaporizer (PTV), and multi-mode injection (MMI) inlets.
  • Supplied clean and ready to use in individually sealed packaging.
  • Highly recommended for trace analysis.