Learn More
Starna Cells Inc SEPTUM CAP SEMI-MICRO BLKWALL
SDP
Catalog No. NC9987751
Encompass_Preferred
Semi-micro Black Walled Quartz Cell w/septum screw top, 10mm
Catalog No. NC9987751 Supplier Starna Cells Inc Supplier No. 9BQ10GL14S
Semi-micro Black Walled Quartz Cell w/septum screw top, 10mm