Learn More
METTLER TOLEDO 3P INDUSTRIAL SEPTUM DL32/DL39 12/PK
Catalog No. NC9578249
Encompass_Preferred
Septum is used for C20 and C30 compact coulometric Karl Fischer titrators. Material: Rubber, Dimensions: 22 mm, Color: Brown, Selling Unit: Pack, Qty/Selling Unit: 12.
Catalog No. NC9578249 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51108740
Septum is used for C20 and C30 compact coulometric Karl Fischer titrators. Material: Rubber, Dimensions: 22 mm, Color: Brown, Selling Unit: Pack, Qty/Selling Unit: 12.