Learn More
Chemglass Life Sciences Septum Holder, M18 Thread, for Bio Reactors
SDP
Catalog No. 501173065
Encompass
Septum Holder, M18 Thread, for Bio Reactors. The septum holder is supplied with 1 silicone rubber septa and can be used as a universal addition port for fluids or gas. Replacement septa are available.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-117-3065 Supplier Chemglass Life Sciences Supplier No. CLS138010
Septum Holder, M18 Thread, for Bio Reactors. The septum holder is supplied with 1 silicone rubber septa and can be used as a universal addition port for fluids or gas. Replacement septa are available.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.