Learn More
MERLIN INSTRUMENT CO SEPTUM MICROSEAL HI PRESSURE
Catalog No. NC9479891
Encompass
Contents: 1 General Purpose Microseal. Requires 23 ga cone tip needle and Merlin Microseal Nut. Inlet pressure: 3-100 psi. Inlet temperature: 50 - 400 C. The Merlin Microseal is a long-lasting microvalve alternative to GC septa.
Catalog No. NC9479891 Supplier MERLIN INSTRUMENT CO Supplier No. 410
Contents: 1 General Purpose Microseal. Requires 23 ga cone tip needle and Merlin Microseal Nut. Inlet pressure: 3-100 psi. Inlet temperature: 50 - 400 C. The Merlin Microseal is a long-lasting microvalve alternative to GC septa.