Learn More
Starna Cells Inc SEPTUM ONLY INSERTS
SDP
Catalog No. NC9776706
Encompass_Preferred
Septum Insert for GL14 Caps. 12.6 dia x 2.4mm thick
Catalog No. NC9776706 Supplier Starna Cells Inc Supplier No. GL14/SI
Septum Insert for GL14 Caps. 12.6 dia x 2.4mm thick