Learn More
Waters Corp Waters Corporation Septum, PTFE, 15 x 45 mm
Catalog No. 50785891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PTFE septum acts as a barrier between a vial and the outside environment, protecting the contents from contaminants. Each septum is thoroughly tested for cleanliness to prevent contamination during sample preparation.
  • Provides optimal conditions for sample injection needles, allowing penetration without introducing contaminants.
  • Protects compounds from contaminants in the air or lab.
  • Made from PTFE (Polytetrafluoroethylene).
  • Dimensions: 15 x 45 mm.
  • Units per package: 144.
  • Designed for sample preparation applications.
Catalog No. 50-785-891 Supplier Waters Corp Supplier No. WAT072714
This PTFE septum acts as a barrier between a vial and the outside environment, protecting the contents from contaminants. Each septum is thoroughly tested for cleanliness to prevent contamination during sample preparation.
  • Provides optimal conditions for sample injection needles, allowing penetration without introducing contaminants.
  • Protects compounds from contaminants in the air or lab.
  • Made from PTFE (Polytetrafluoroethylene).
  • Dimensions: 15 x 45 mm.
  • Units per package: 144.
  • Designed for sample preparation applications.