Learn More
Bellco Glass, Inc. Septum Stopper,13mm, Hungate Style, Black Butyl Rubber
SDP
Catalog No. 501945336
Encompass_Preferred
Hungate stopper only for use with hungate tubes,2047-00125. Size: 13mm Dimensions: 13mm @ top, taper to 10mm x 5mm thick. MADE IN THE USA
Catalog No. 50-194-5336 Supplier Bellco Glass, Inc. Supplier No. 204711600
Hungate stopper only for use with hungate tubes,2047-00125. Size: 13mm Dimensions: 13mm @ top, taper to 10mm x 5mm thick. MADE IN THE USA