Learn More
Metrohm USA SEPTUM STOPPER
SDP
Supplier:  Metrohm USA 62730020
Encompass
With E.301.0041 O-ring. For 6.1448.010 Septum and 6.1414.030 Titration vessel lid. Used for KF applications.
Catalog No. NC0136783