Learn More
Bellco Glass, Inc. Septum Stopper, 20mm Blue Butyl Rubber
SDP
Catalog No. 501945347
Encompass_Preferred
Manufactured from blue Chlorobutyl. The septum stopper can withstand autoclaving 160 degree Celsius at normal autoclaving times 12-20 minutes. Autoclaving: no higher than 160 degree Celsius. FOR 2048-00150 - ANAEROBIC TUBES. DIMENSIONS: 19.5mm @ TOP, 14mm LONG, 14.5-13.5mm TAPER. MADE IN THE USA. Size: 20mm
Catalog No. 50-194-5347 Supplier Bellco Glass, Inc. Supplier No. 204811800A
Manufactured from blue Chlorobutyl. The septum stopper can withstand autoclaving 160 degree Celsius at normal autoclaving times 12-20 minutes. Autoclaving: no higher than 160 degree Celsius. FOR 2048-00150 - ANAEROBIC TUBES. DIMENSIONS: 19.5mm @ TOP, 14mm LONG, 14.5-13.5mm TAPER. MADE IN THE USA. Size: 20mm