Learn More
Starna Cells Inc SEPTUM TOP QUARTZ CELL
SDP
Catalog No. NC9776704
Encompass_Preferred
Fluorometer Cells,Standard Rectangular,Screw Cap with septum,Window material Spectrosil Quartz or equivalent,useable range:170 to 2700nm,Path Length mm:10,Exterior WxLxH mm:12.5 x 12.5 x 68,Interior W x L mm:10 x 10,Nominal Volume ml:3.5,Nu
Catalog No. NC9776704 Supplier Starna Cells Inc Supplier No. 3Q10GL14S
Fluorometer Cells,Standard Rectangular,Screw Cap with septum,Window material Spectrosil Quartz or equivalent,useable range:170 to 2700nm,Path Length mm:10,Exterior WxLxH mm:12.5 x 12.5 x 68,Interior W x L mm:10 x 10,Nominal Volume ml:3.5,Nu