Learn More
PRO LAB SUPPLY CORP SPL Septum Tube 15ml, (Tube PP/ Cap HDPE)
SDP
Catalog No. NC2877328
Encompass_Preferred
SPL Septum Tube 15ml, (Tube PP/ Cap HDPE), Septum (Silicone/PTFE) RCF Rating 13,000 x g, Sterile Case of 25. Tight sealing screw cap. Silicone septum with PTFE coating. External graduations with marking area. Provided in rack. Packed in zipper bags. Non-Cytotoxic, Non-Pyrogenic, DNase/RNase - free, Human DNA - free
Catalog No. NC2877328 Supplier PRO LAB SUPPLY CORP Supplier No. 50715
SPL Septum Tube 15ml, (Tube PP/ Cap HDPE), Septum (Silicone/PTFE) RCF Rating 13,000 x g, Sterile Case of 25. Tight sealing screw cap. Silicone septum with PTFE coating. External graduations with marking area. Provided in rack. Packed in zipper bags. Non-Cytotoxic, Non-Pyrogenic, DNase/RNase - free, Human DNA - free