Learn More
Takara Bio SeqAmp™ CB PCR Buffer | 50 Rxns
SDP
Catalog No. NC1384333
Encompass
SeqAmp™ CB PCR Buffer is designed for improved compatibility with DNA purification using Agencourt AMPure beads after amplification with SeqAmp DNA Polymerase and works with SMARTer next-generation sequencing kits.
Catalog No. NC1384333 Supplier Takara Bio Supplier No. 638526
SeqAmp™ CB PCR Buffer is designed for improved compatibility with DNA purification using Agencourt AMPure beads after amplification with SeqAmp DNA Polymerase and works with SMARTer next-generation sequencing kits.