Learn More
Sigma Aldrich Fine Chemicals Biosciences SeqPlex RNA Amplification
Catalog No. 502914686
Encompass
The SeqPlex RNA Amplification Kit for whole transcriptome amplification (WTA) is designed to facilitate next-generation sequencing (NGS) from small quantities or from degraded/highly fragmented RNA (e.g. RNA from formalin-fixed paraffin-embedded (ffPE) tissue samples). The SeqPlex kit allows the user to pre-amplify these and other small quantity/highly fragmented RNA samples for input into a NGS workflow.
Catalog No. 50-291-4686 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SEQR10RXN
The SeqPlex RNA Amplification Kit for whole transcriptome amplification (WTA) is designed to facilitate next-generation sequencing (NGS) from small quantities or from degraded/highly fragmented RNA (e.g. RNA from formalin-fixed paraffin-embedded (ffPE) tissue samples). The SeqPlex kit allows the user to pre-amplify these and other small quantity/highly fragmented RNA samples for input into a NGS workflow.