Learn More
National Diagnostics SEQUAGEL BUFFER 200 ML
SDP
Catalog No. 5089990072
Encompass
SequaGel Buffer, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 200 ML
Catalog No. 50-899-90072 Supplier National Diagnostics Supplier No. EC835200ML
Add to Cart
Add to Cart
SequaGel Buffer, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 200 ML