Learn More
National Diagnostics SEQUAGEL CONCENTRATE 450 ML
SDP
Catalog No. 5089990068
Encompass
SequaGel Concentrate, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 450 ML
Catalog No. 50-899-90068 Supplier National Diagnostics Supplier No. EC830450ML
SequaGel Concentrate, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 450 ML