Learn More
National Diagnostics SEQUAGEL DILUENT 1 LT
SDP
Catalog No. 5089990077
Encompass
SequaGel Diluent, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 1 LT
Catalog No. 50-899-90077 Supplier National Diagnostics Supplier No. EC8401LTR
SequaGel Diluent, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 1 LT