Learn More
National Diagnostics SEQUAGEL DILUENT 450 ML
SDP
Catalog No. 5089990078
Encompass
SequaGel Diluent, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 450 ML
Catalog No. 50-899-90078 Supplier National Diagnostics Supplier No. EC840450ML
SequaGel Diluent, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 450 ML