Learn More
National Diagnostics SEQUAGEL SEQUENCING SYSTEM 1 L
SDP
Catalog No. 5089990069
Encompass
SequaGel Sequencing System, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 1 LT
Catalog No. 50-899-90069 Supplier National Diagnostics Supplier No. EC8331LTR
SequaGel Sequencing System, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 1 LT