Learn More
National Diagnostics SEQUAGEL SEQUENCING SYSTEM 2.2
SDP
Catalog No. 5089990070
Encompass
SequaGel Sequencing System, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 2.2 LT
Catalog No. 50-899-90070 Supplier National Diagnostics Supplier No. EC8332.2LTR
SequaGel Sequencing System, System consists of SequaGel Concentrate, SequaGel Diluent, and SequaGel Buffer for casting 19:1 denaturing DNA gels of a varying percentage, 2.2 LT